Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290258.1 | NP_001277187.1 | 10 | 1417 | ||
NM_080928.4 | NP_563616.3 | 8 | 1202 | |||
XM_005250146.4 | XP_005250203.1 | 10 | 1537 | |||
XM_011515816.2 | XP_011514118.1 | 8 | 1198 | |||
XM_011515817.2 | XP_011514119.1 | 11 | 1598 | |||
XM_011515818.2 | XP_011514120.1 | 11 | 1635 | |||
XM_011515819.2 | XP_011514121.1 | 11 | 1580 | |||
XM_011515820.2 | XP_011514122.1 | 9 | 1412 | |||
XM_011515821.2 | XP_011514123.1 | 7 | 1039 | |||
XM_017011756.1 | XP_016867245.1 | 11 | 2483 | |||
XM_017011757.1 | XP_016867246.1 | 11 | 1713 | |||
XM_017011758.1 | XP_016867247.1 | 4 | 761 | |||
GenBank mRNA | AF403033.1 | - | 3 | 664 | ||
AF428257.1 | - | 9 | 1113 | |||
AK125360.1 | - | 10 | 1417 | |||
CR749595.1 | - | 5 | 944 |