Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098483.2 | NP_001091953.1 | 6 | 649 | ||
NM_001301827.1 | NP_001288756.1 | 5 | 580 | |||
NM_001301828.1 | NP_001288757.1 | 6 | 856 | |||
NM_198472.2 | NP_940874.2 | 6 | 718 | |||
XM_011539670.2 | XP_011537972.1 | 6 | 721 | |||
XM_011539671.2 | XP_011537973.1 | 6 | 688 | |||
XM_011539672.2 | XP_011537974.1 | 6 | 688 | |||
XM_017016094.1 | XP_016871583.1 | 6 | 919 | |||
GenBank mRNA | AK129527.1 | - | 6 | 718 | ||
BM009909.1 | - | 5 | 513 | |||
CN368132.1 | - | 1 | 155 |