Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001791.2 | NP_001001791.2 | 5 | 564 | ||
GenBank mRNA | AK092517.1 | - | 2 | 209 | ||
BC049374.1 | - | 5 | 564 | 5266108 |