Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNIP2 SIRNA Assays ›
Gene Symbol:
KCNIP2
KCNIP2-AS1
Gene Name:
potassium voltage-gated channel interacting protein 2
KCNIP2 antisense RNA 1
Gene Aliases:
KCHIP2
-
Chromosome Location:
Chr.10: 101825971 - 101843920 on Build GRCh38
Chromosome Location:
Chr.10: 101819068 - 101828889 on Build GRCh38
UniGene ID:
Hs.97044
Hs.568881
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
21620
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNIP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014591.4
NP_055406.2
5
780
NM_173191.2
NP_775283.1
5
735
NM_173192.2
NP_775284.1
4
681
NM_173193.2
NP_775285.1
3
606
NM_173194.2
NP_775286.1
3
262
NM_173195.2
NP_775287.1
3
585
NM_173197.2
NP_775289.1
3
585
XM_005269729.2
XP_005269786.1
4
583
XM_005269730.2
XP_005269787.1
3
540
XM_006717812.2
XP_006717875.1
5
637
XM_011539731.2
XP_011538033.1
3
277
XM_017016161.1
XP_016871650.1
3
332
GenBank mRNA
AB044584.1
-
5
383
AB044585.1
-
3
233
AF199598.1
-
4
329
AF295076.1
-
3
233
AF295530.1
-
3
254
AF347114.1
-
3
233
AF367018.1
-
5
428
AF367019.1
-
3
233
AF367020.1
-
2
175
AF367021.1
-
5
383
AK027347.1
-
3
253
AK055089.1
-
5
659
AK094668.1
-
7
787
AK125382.1
-
3
1292
AK295324.1
-
1
216
AK301433.1
-
3
254
AK309368.1
-
3
461
AK314536.1
-
3
466
AK315042.1
-
4
561
AL136722.1
-
3
280
AY026328.1
-
3
233
AY302141.1
-
3
233
BC034685.1
-
4
544
4155871
BG911444.1
-
1
198
DQ148480.1
-
5
383
DQ148481.1
-
4
329
DQ148482.1
-
3
233
DQ148483.1
-
3
248
DQ148484.1
-
2
175
DQ862463.1
-
3
233
Gene Symbol: KCNIP2-AS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AI125777.1
-
1
234
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_020094, NM_001033961, NM_020095, XM_008760448, XM_008760483, and others.This siRNA also targets the following transcripts in Mouse: AB044571, AB044570.
Gene Ontology Categories:
Function(s)
Process(es)
detection of calcium ion
potassium ion transport
muscle contraction
signal transduction
chemical synaptic transmission
regulation of heart contraction
clustering of voltage-gated potassium channels
cardiac conduction
potassium ion export
potassium ion transmembrane transport
membrane repolarization
regulation of potassium ion transmembrane transport
regulation of cation channel activity
A-type (transient outward) potassium channel activity
calcium ion binding
protein binding
potassium channel regulator activity
ion channel binding
ER retention sequence binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline