Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256841.1 | NP_001243770.1 | 5 | 674 | ||
NM_007261.3 | NP_009192.2 | 6 | 1013 | |||
XM_005256992.4 | XP_005257049.1 | 6 | 942 | |||
XM_006721656.3 | XP_006721719.1 | 6 | 1050 | |||
GenBank mRNA | AF020314.1 | - | 6 | 778 | ||
AJ010101.1 | - | 6 | 721 | |||
AJ010102.1 | - | 5 | 382 | |||
AJ010103.1 | - | 3 | 133 | |||
AJ224864.1 | - | 6 | 994 | |||
AJ238323.1 | - | 6 | 721 | |||
AK309631.1 | - | 7 | 1078 | |||
AK309636.1 | - | 5 | 1119 | |||
BC032352.1 | - | 6 | 978 | 5218501 |