Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256841.1 | NP_001243770.1 | 6 | 775 | ||
NM_007261.3 | NP_009192.2 | 7 | 1114 | |||
XM_005256990.2 | XP_005257047.1 | 6 | 1027 | |||
XM_005256991.4 | XP_005257048.1 | 5 | 991 | |||
XM_005256992.4 | XP_005257049.1 | 7 | 1043 | |||
XM_006721656.3 | XP_006721719.1 | 7 | 1151 | |||
XM_011524254.2 | XP_011522556.1 | 5 | 730 | |||
GenBank mRNA | AF020314.1 | - | 7 | 879 | ||
AF161346.1 | - | 5 | 391 | |||
AJ010101.1 | - | 7 | 822 | |||
AJ010102.1 | - | 6 | 483 | |||
AJ010103.1 | - | 4 | 234 | |||
AJ224864.1 | - | 7 | 1095 | |||
AJ238323.1 | - | 7 | 822 | |||
AK309631.1 | - | 8 | 1179 | |||
AK309636.1 | - | 6 | 1220 | |||
BC032352.1 | - | 7 | 1079 | 5218501 |