Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182511.3 | NP_872317.1 | 5 | 1451 | ||
XM_006722394.3 | XP_006722457.1 | 7 | 1798 | |||
XM_011525824.2 | XP_011524126.1 | 7 | 1801 | |||
XM_017025559.1 | XP_016881048.1 | 7 | 2302 | |||
XM_017025560.1 | XP_016881049.1 | 4 | 963 | |||
GenBank mRNA | AK125422.1 | - | 2 | 1897 | ||
AK291364.1 | - | 4 | 963 | |||
AY359074.1 | - | 5 | 1291 | |||
BC035789.1 | - | 4 | 953 | 5727802 |