Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026838.1 | 4 | 691 | |||
NR_026839.1 | 3 | 594 | ||||
NR_026840.1 | 4 | 452 | ||||
NR_026841.1 | 4 | 434 | ||||
NR_026842.1 | 3 | 337 | ||||
GenBank mRNA | AF321193.1 | - | 4 | 409 | ||
AF426256.1 | - | 4 | 357 | |||
AJ001904.1 | - | 4 | 394 | |||
AJ297914.2 | - | 4 | 412 | |||
AJ301615.2 | - | 3 | 315 | |||
AJ305085.2 | - | 3 | 572 | |||
AJ306839.2 | - | 4 | 430 | |||
AJ306840.2 | - | 4 | 484 | |||
AJ783421.1 | - | 4 | 669 | |||
BC015981.1 | - | 3 | 319 | |||
BC029353.1 | - | 3 | 594 | |||
BC070229.1 | - | 3 | 296 | |||
BC117304.1 | - | 4 | 366 | |||
BC117330.1 | - | 4 | 366 | |||
BC143995.1 | - | 4 | 384 | |||
BG189423.1 | - | 1 | 89 |