Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIP1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003959.2 | NP_003950.1 | 32 | 3879 | ||
XM_011538963.1 | XP_011537265.1 | 32 | 4182 | |||
GenBank mRNA | AB013384.1 | - | 26 | 3232 | ||
AB014555.1 | - | 32 | 3813 | |||
AK307446.1 | - | 9 | 1456 | |||
AW962010.1 | - | 2 | 491 | |||
CF454957.1 | - | 381 |