Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033063.1 | NP_149052.1 | 2, 3 | 1192 | ||
NM_207577.1 | NP_997460.1 | 2, 3 | 1192 | |||
XM_017017757.1 | XP_016873246.1 | 3, 4 | 361 | |||
GenBank mRNA | AB058781.1 | - | 2, 3 | 1192 | ||
AK123340.1 | - | 2, 3 | 300 | |||
BC063860.1 | - | 2, 3 | 858 | |||
BC139780.1 | - | 3, 4 | 431 | 40122938 | ||
BC150254.1 | - | 2, 3 | 1192 | 9007181 |