Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033063.1 | NP_149052.1 | 2 | 1121 | ||
NM_207577.1 | NP_997460.1 | 2 | 1121 | |||
XM_006718556.3 | XP_006718619.1 | 2 | 3760 | |||
XM_011545039.2 | XP_011543341.1 | 2 | 3761 | |||
XM_017017755.1 | XP_016873244.1 | 2 | 3660 | |||
XM_017017756.1 | XP_016873245.1 | 2 | 223 | |||
XM_017017757.1 | XP_016873246.1 | 3 | 290 | |||
XM_017017758.1 | XP_016873247.1 | 2 | 257 | |||
GenBank mRNA | AB058781.1 | - | 2 | 1121 | ||
AK123340.1 | - | 2 | 229 | |||
BC063860.1 | - | 2 | 787 | |||
BC139780.1 | - | 3 | 360 | 40122938 | ||
BC150254.1 | - | 2 | 1121 | 9007181 |