Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001655.2 | NP_001001655.1 | 3 | 647 | ||
NM_001145374.1 | NP_001138846.1 | 3 | 797 | |||
NM_001145375.1 | NP_001138847.1 | 3 | 544 | |||
XM_005253835.4 | XP_005253892.1 | 3 | 813 | |||
GenBank mRNA | AY754389.1 | - | 3 | 632 | ||
BC070489.1 | - | 3 | 599 | 6048174 | ||
BM790974.1 | - | 3 | 526 |