Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001668.3 | NP_001001668.3 | 6 | 2664 | ||
XM_017026801.1 | XP_016882290.1 | 7 | 2738 | |||
GenBank mRNA | AK129686.1 | - | 1 | 1428 | ||
AY484591.1 | - | 6 | 2180 | |||
BC136762.1 | - | 6 | 2176 | 9020752 | ||
BX641145.1 | - | 1 | 1627 |