Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014630.2 | NP_055445.2 | 4 | 572 | ||
XM_005254996.3 | XP_005255053.1 | 3 | 473 | |||
XM_011522246.2 | XP_011520548.1 | 4 | 642 | |||
XM_011522247.2 | XP_011520549.1 | 3 | 645 | |||
XM_011522248.2 | XP_011520550.1 | 4 | 603 | |||
XM_017022734.1 | XP_016878223.1 | 2 | 353 | |||
GenBank mRNA | BC094688.1 | - | 4 | 524 | 30397050 | |
BC112232.1 | - | 2 | 306 | 8327700 | ||
BC112234.1 | - | 2 | 307 | 8327702 | ||
BC143600.1 | - | 2 | 308 | |||
D86966.1 | - | 4 | 806 |