Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3PXD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014631.2 | NP_055446.2 | 10 | 910 | ||
XM_005270294.4 | XP_005270351.1 | 11 | 1043 | |||
XM_005270295.4 | XP_005270352.1 | 10 | 997 | |||
XM_005270297.3 | XP_005270354.1 | 5 | 2274 | |||
XM_005270298.4 | XP_005270355.1 | 5 | 1077 | |||
XM_006718079.3 | XP_006718142.1 | 12 | 1140 | |||
XM_006718080.3 | XP_006718143.1 | 6 | 1171 | |||
XM_011540385.2 | XP_011538687.1 | 11 | 1094 | |||
XM_011540386.2 | XP_011538688.1 | 6 | 2370 | |||
XM_011540387.2 | XP_011538689.1 | 2 | 4016 | |||
XM_017016969.1 | XP_016872458.1 | 11 | 1061 | |||
XM_017016970.1 | XP_016872459.1 | 10 | 877 | |||
GenBank mRNA | AB007878.1 | - | 8 | 660 | ||
AK056469.1 | - | 9 | 633 | |||
AK294454.1 | - | 5 | 450 | |||
AK294736.1 | - | 5 | 488 | |||
AK295314.1 | - | 5 | 527 | |||
AK295660.1 | - | 5 | 554 | |||
AK302015.1 | - | 8 | 666 | |||
AK304828.1 | - | 5 | 466 | |||
AK315983.1 | - | 5 | 493 |