Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042385.2 | NP_001035844.1 | 4 | 583 | ||
NM_001042386.2 | NP_001035845.1 | 4 | 583 | |||
NM_001171603.1 | NP_001165074.1 | 5 | 923 | |||
NM_001171606.1 | NP_001165077.1 | 5 | 1054 | |||
NM_001171613.1 | NP_001165084.1 | 4 | 512 | |||
NM_001171617.1 | NP_001165088.1 | 4 | 669 | |||
NM_006036.4 | NP_006027.2 | 4 | 1508 | |||
XM_011533198.1 | XP_011531500.1 | 5 | 1010 | |||
XM_011533200.1 | XP_011531502.1 | 5 | 662 | |||
XM_011533202.1 | XP_011531504.1 | 4 | 462 | |||
XM_017005384.1 | XP_016860873.1 | 5 | 1648 | |||
XM_017005385.1 | XP_016860874.1 | 5 | 973 | |||
GenBank mRNA | AB007896.1 | - | 4 | 435 | ||
AK025649.1 | - | 4 | 355 | |||
AK131463.1 | - | 5 | 866 | |||
BC151236.1 | - | 5 | 987 | 8860445 | ||
BG717492.1 | - | 4 | 617 | |||
DA246860.1 | - | 4 | 512 | |||
DQ023503.1 | - | 4 | 1508 | |||
DQ023504.1 | - | 4 | 1508 | |||
DQ023505.1 | - | 4 | 1508 | |||
DQ023506.1 | - | 4 | 583 | |||
DQ023507.1 | - | 4 | 583 |