Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPECC1L-ADORA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103546.1 | 4 | 516 | |||
GenBank mRNA | AB209022.1 | - | 4 | 435 |
Gene Symbol: DGCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_110533.1 | 5 | 766 | |||
GenBank mRNA | X91348.1 | - | LIT1227 DGCR5 |
5 | 738 |
Gene Symbol: SPECC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145468.3 | NP_001138940.3 | 3 | 412 | ||
NM_001254732.2 | NP_001241661.2 | 3 | 412 | |||
NM_015330.4 | NP_056145.4 | 4 | 516 | |||
GenBank mRNA | AK097129.1 | - | 3 | 400 | ||
AK301482.1 | - | 3 | 347 | |||
AL832425.1 | - | 4 | 435 | |||
AY884293.1 | - | 3 | 340 | |||
BC005093.1 | - | 4 | 435 | |||
BC136471.1 | - | 3 | 282 | 9020460 | ||
DA385554.1 | - | 3 | 406 |