Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPECC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145468.3 | NP_001138940.3 | 16 | 6085 | ||
NM_001254732.2 | NP_001241661.2 | 15 | 5968 | |||
NM_001254733.1 | NP_001241662.1 | 9 | 3624 | |||
NM_015330.4 | NP_056145.4 | 17 | 6189 | |||
GenBank mRNA | AB002374.1 | - | 13 | 5414 | ||
AK025531.1 | - | 13 | 4493 | |||
AL832425.1 | - | 17 | 6108 | |||
BC021132.1 | - | 1 | 1549 | |||
BC041571.1 | - | 1 | 1639 | |||
BQ929263.1 | - | 1 | 536 |