Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023570.3 | NP_001018864.2 | 5 | 512 | ||
NM_001023571.3 | NP_001018865.2 | 5 | 512 | |||
NM_001319107.1 | NP_001306036.1 | 5 | 511 | |||
NR_134968.1 | 5 | 511 | ||||
XM_005247911.3 | XP_005247968.1 | 5 | 511 | |||
XM_005247912.2 | XP_005247969.1 | 4 | 325 | |||
XM_011513335.2 | XP_011511637.1 | 4 | 325 | |||
XM_017007537.1 | XP_016863026.1 | 5 | 488 | |||
XM_017007539.1 | XP_016863028.1 | 5 | 498 | |||
GenBank mRNA | AB062481.1 | - | 5 | 505 | ||
AY714228.1 | - | 5 | 506 | |||
AY964667.1 | - | 5 | 462 | |||
AY964668.1 | - | 5 | 450 | |||
BC005806.2 | - | 5 | 453 | 3834623 | ||
BG721354.1 | - | 516 | ||||
BQ068103.1 | - | 499 | ||||
D25278.1 | - | 5 | 453 |