Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290187.1 | NP_001277116.1 | 11 | 1579 | ||
NM_032534.3 | NP_115923.2 | 11 | 1878 | |||
XM_011516636.2 | XP_011514938.1 | 12 | 1806 | |||
XM_011516637.2 | XP_011514939.1 | 12 | 1803 | |||
XM_011516638.2 | XP_011514940.1 | 12 | 1762 | |||
XM_011516639.2 | XP_011514941.1 | 12 | 1758 | |||
XM_011516640.2 | XP_011514942.1 | 12 | 1755 | |||
XM_011516641.2 | XP_011514943.1 | 11 | 1694 | |||
XM_011516642.2 | XP_011514944.1 | 11 | 1846 | |||
XM_011516643.2 | XP_011514945.1 | 11 | 1707 | |||
GenBank mRNA | AB058765.2 | - | 11 | 1832 | ||
AL831935.1 | - | 9 | 1303 | |||
BC033229.1 | - | 4 | 533 | |||
BC041587.1 | - | 4 | 533 | |||
BC142722.1 | - | 11 | 1579 | 40148879 | ||
BC146667.1 | - | 11 | 1579 | 40148146 | ||
BC150330.1 | - | 11 | 1843 | 8860497 | ||
BC152409.1 | - | 11 | 1832 | 8862512 |