Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARMIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138360.3 | NP_612369.3 | 39 | 4221 | ||
XM_011537300.2 | XP_011535602.1 | 39 | 4012 | |||
XM_017021751.1 | XP_016877240.1 | 36 | 3913 | |||
GenBank mRNA | AK074167.1 | - | 23 | 3938 | ||
AL834197.1 | - | 10 | 2289 | |||
BC008134.2 | - | 6 | 668 |