Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284292.1 | NP_001271221.1 | 8 | 3818 | ||
NM_001284293.1 | NP_001271222.1 | 7 | 3724 | |||
NM_175741.2 | NP_786883.1 | 8 | 3903 | |||
GenBank mRNA | AF482429.1 | - | 7 | 3507 | ||
AK302656.1 | - | 8 | 3818 | |||
AK302680.1 | - | 7 | 3461 | |||
AL137416.1 | - | 1 | 686 | |||
BC114518.1 | - | 7 | 3479 | 40036588 | ||
BC114567.1 | - | 7 | 3474 | 40036589 |