Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256167.1 | NP_001243096.1 | 3 | 946 | ||
NM_001256168.1 | NP_001243097.1 | 3 | 936 | |||
NM_175888.3 | NP_787084.1 | 5 | 1187 | |||
XM_005265011.3 | XP_005265068.1 | 5 | 4449 | |||
XM_005265012.3 | XP_005265069.1 | 5 | 4374 | |||
XM_017006064.1 | XP_016861553.1 | 6 | 4586 | |||
XM_017006065.1 | XP_016861554.1 | 4 | 4273 | |||
XM_017006066.1 | XP_016861555.1 | 6 | 4490 | |||
XM_017006067.1 | XP_016861556.1 | 4 | 1085 | |||
XM_017006068.1 | XP_016861557.1 | 3 | 985 | |||
XM_017006069.1 | XP_016861558.1 | 3 | 4368 | |||
GenBank mRNA | AK093599.1 | - | 3 | 945 | ||
AK131232.1 | - | 5 | 1187 |