Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014646.2 | NP_055461.1 | 3 | 517 | ||
XM_005258177.4 | XP_005258234.1 | 3 | 587 | |||
XM_005258178.3 | XP_005258235.1 | 3 | 516 | |||
XM_005258179.4 | XP_005258236.1 | 3 | 450 | |||
XM_017026098.1 | XP_016881587.1 | 4 | 606 | |||
XM_017026099.1 | XP_016881588.1 | 3 | 377 | |||
GenBank mRNA | BC152448.1 | - | 3 | 517 | 8862579 | |
D87436.1 | - | 3 | 517 |