Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182532.2 | NP_872338.1 | 3 | 1038 | ||
XM_005270586.4 | XP_005270643.1 | 3 | 676 | |||
XM_011540911.2 | XP_011539213.1 | 4 | 790 | |||
XM_011540912.2 | XP_011539214.1 | 3 | 1566 | |||
GenBank mRNA | BC029775.1 | - | 3 | 910 | 5186268 |