Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144939.1 | NP_001138411.1 | 3 | 777 | ||
NM_001144940.1 | NP_001138412.1 | 3 | 721 | |||
NM_001144941.1 | NP_001138413.1 | 2 | 625 | |||
NM_182566.2 | NP_872372.1 | 3 | 741 | |||
GenBank mRNA | AF521892.1 | - | 3 | 746 | ||
AY359099.1 | - | 3 | 671 | |||
BC038467.1 | - | 2 | 624 | |||
BC104194.2 | - | 3 | 536 | 40031204 | ||
BC104195.1 | - | 3 | 709 | 40031205 | ||
BI836994.1 | - | 2 | 625 |