Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198546.1 | NP_940948.1 | 13 | 2001 | ||
XM_011541407.2 | XP_011539709.1 | 14 | 2513 | |||
XM_017001224.1 | XP_016856713.1 | 10 | 1561 | |||
GenBank mRNA | AY327405.1 | - | 13 | 2001 |