Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10G8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004464.1 | NP_001004464.1 | 1 | 580 | ||
GenBank mRNA | BC136917.1 | - | 1 | 613 | 9020908 | |
BC136920.1 | - | 1 | 613 | 9020911 |