Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004297.2 | NP_001004297.2 | 4 | 1644 | ||
XM_011540147.2 | XP_011538449.1 | 3 | 2111 | |||
XM_017016632.1 | XP_016872121.1 | 4 | 5030 | |||
XM_017016633.1 | XP_016872122.1 | 3 | 5172 | |||
XM_017016634.1 | XP_016872123.1 | 3 | 5417 | |||
GenBank mRNA | AK131555.1 | - | 4 | 1644 |