Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272005.1 | NP_001258934.1 | 4, 5 | 748 | ||
NM_178233.2 | NP_839947.1 | 4, 5 | 682 | |||
XM_011524744.2 | XP_011523046.1 | 4, 5 | 678 | |||
GenBank mRNA | AB593153.1 | - | 4, 5 | 668 | ||
BC143799.1 | - | 4, 5 | 831 |