Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIMBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015347.4 | NP_056162.4 | 8 | 1155 | ||
XM_011538102.1 | XP_011536404.1 | 9 | 1256 | |||
XM_011538103.2 | XP_011536405.1 | 11 | 1718 | |||
XM_011538104.2 | XP_011536406.1 | 11 | 1533 | |||
XM_011538105.2 | XP_011536407.1 | 11 | 1707 | |||
XM_011538106.2 | XP_011536408.1 | 11 | 1707 | |||
XM_011538107.2 | XP_011536409.1 | 11 | 1714 | |||
XM_011538108.2 | XP_011536410.1 | 11 | 1713 | |||
XM_017019105.1 | XP_016874594.1 | 11 | 1731 | |||
XM_017019106.1 | XP_016874595.1 | 11 | 1707 | |||
XM_017019107.1 | XP_016874596.1 | 11 | 1529 | |||
XM_017019108.1 | XP_016874597.1 | 11 | 1712 | |||
XM_017019109.1 | XP_016874598.1 | 9 | 1256 | |||
XM_017019110.1 | XP_016874599.1 | 11 | 1529 | |||
XM_017019111.1 | XP_016874600.1 | 11 | 1529 | |||
XM_017019112.1 | XP_016874601.1 | 11 | 1751 | |||
GenBank mRNA | AB002316.1 | - | 8 | 1155 | ||
AY699979.1 | - | 5 | 846 | |||
BC007632.1 | - | 12 | 2042 | 3507574 |