Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198274.3 | NP_938015.1 | 10 | 1823 | ||
XM_005264156.2 | XP_005264213.1 | 9 | 1739 | |||
GenBank mRNA | AF086123.1 | - | 2 | 442 | ||
AK091724.1 | - | 10 | 1741 | |||
AL832035.1 | - | 10 | 1740 | |||
AY518933.1 | - | 10 | 1720 | |||
AY518934.1 | - | 6 | 1089 | |||
BC126191.1 | - | 10 | 1823 | 8991907 | ||
BX647878.1 | - | 10 | 1742 |