Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142649.1 | NP_001136121.1 | 22 | 3043 | ||
NM_213599.2 | NP_998764.1 | 22 | 3046 | |||
XM_005252820.3 | XP_005252877.2 | 21 | 3150 | |||
XM_005252821.3 | XP_005252878.2 | 21 | 3147 | |||
XM_005252822.4 | XP_005252879.1 | 22 | 2931 | |||
XM_011519949.2 | XP_011518251.1 | 20 | 3099 | |||
GenBank mRNA | AB125267.1 | - | 22 | 3046 | ||
AK125046.1 | - | 7 | 947 | |||
AL832374.1 | - | 22 | 3068 | |||
AL833271.1 | - | 22 | 3067 |