Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTYH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025250.2 | NP_079526.1 | 14 | 4608 | ||
XM_017012656.1 | XP_016868145.1 | 13 | 4480 | |||
GenBank mRNA | AB051478.2 | - | 14 | 4603 | ||
AB162931.1 | - | 14 | 4433 | |||
AK074158.1 | - | 10 | 3915 | |||
AK124608.1 | - | 11 | 3922 | |||
AL833825.1 | - | 1 | 2903 | |||
BC069027.1 | - | 1 | 1751 | |||
BC152447.1 | - | 14 | 4603 | 8862578 |