Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015051.2 | NP_055866.1 | 5 | 546 | ||
GenBank mRNA | AB011145.1 | - | 5 | 470 | ||
AJ344330.2 | - | 5 | 457 | |||
AK075024.1 | - | 5 | 534 | |||
AY359048.1 | - | 5 | 470 | |||
BC005374.1 | - | 5 | 507 | 3934201 | ||
BQ881761.1 | - | 5 | 546 | |||
BX648519.1 | - | 6 | 637 |