Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160183.2 | NP_001153655.1 | 5 | 934 | ||
NM_001271637.1 | NP_001258566.1 | 4 | 801 | |||
NM_001271638.1 | NP_001258567.1 | 4 | 808 | |||
NM_001271639.1 | NP_001258568.1 | 4 | 797 | |||
NM_001271640.1 | NP_001258569.1 | 4 | 856 | |||
NM_001271649.1 | NP_001258578.1 | 2 | 592 | |||
NM_006524.3 | NP_006515.2 | 3 | 719 | |||
NR_073391.1 | 5 | 901 | ||||
GenBank mRNA | AK055644.1 | - | 4 | 761 | ||
AK298230.1 | - | 3 | 684 | |||
BC131762.1 | - | 3 | 683 | 40121140 | ||
BC141847.1 | - | 4 | 768 | |||
U09847.1 | - | 1 | 384 |