Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114107.4 | NP_001107579.1 | 1, 2 | 221 | ||
NM_001257962.1 | NP_001244891.1 | 1, 2 | 221 | |||
NM_014476.5 | NP_055291.2 | 1, 2 | 221 | |||
NR_047562.1 | 1, 2 | 221 | ||||
GenBank mRNA | AF002280.1 | - | 1, 2 | 143 | ||
AF002282.1 | - | 1, 2 | 143 | |||
AF039018.1 | - | 1, 2 | 167 | |||
AK129863.1 | - | 1, 2 | 90 | |||
AK222966.1 | - | 1, 2 | 125 | |||
AK313253.1 | - | 1, 2 | 170 | |||
BC027870.1 | - | 1, 2 | 148 | 5205215 | ||
BI913867.1 | - | 1, 2 | 221 | |||
BT007341.1 | - | 1, 2 | 97 | |||
C03422.1 | - | 1, 2 | 47 |