Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018263.4 | NP_060733.4 | 12 | 1870 | ||
XM_006712039.3 | XP_006712102.1 | 8 | 3991 | |||
XM_006712040.2 | XP_006712103.1 | 8 | 1277 | |||
XM_011532950.2 | XP_011531252.1 | 11 | 1909 | |||
XM_011532951.2 | XP_011531253.1 | 10 | 1597 | |||
XM_017004429.1 | XP_016859918.1 | 12 | 1700 | |||
XM_017004430.1 | XP_016859919.1 | 9 | 4057 | |||
GenBank mRNA | AB051472.2 | - | 12 | 1870 | ||
AB084279.1 | - | 12 | 1733 | |||
BC042999.2 | - | 8 | 1308 | 5297392 | ||
BX647527.1 | - | 13 | 1943 | |||
BX648218.1 | - | 12 | 1895 | |||
CR933613.1 | - | 12 | 1914 |