Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012227.3 | NP_036359.3 | 5, 6 | 797 | ||
XM_006724868.3 | XP_006724931.1 | 6, 7 | 697 | |||
XM_011545637.2 | XP_011543939.1 | 4, 5 | 402 | |||
GenBank mRNA | AK024630.1 | - | 5, 6 | 736 | ||
AK223412.1 | - | 5, 6 | 742 | |||
AK296003.1 | - | 5, 6 | 458 | |||
AK298260.1 | - | 5, 6 | 544 | |||
BC014636.1 | - | 5, 6 | 470 | 3528598 | ||
BT007360.1 | - | 4, 5 | 426 | |||
Y14391.2 | - | 5, 6 | 755 |