Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004626.2 | NP_004617.2 | 5 | 1401 | ||
XM_005274231.1 | XP_005274288.1 | 6 | 1657 | |||
XM_011545238.1 | XP_011543540.1 | 6 | 1687 | |||
XM_011545239.2 | XP_011543541.1 | 6 | 1895 | |||
XM_011545240.2 | XP_011543542.1 | 7 | 1851 | |||
XM_011545241.2 | XP_011543543.1 | 5 | 1139 | |||
XM_017018241.1 | XP_016873730.1 | 4 | 2159 | |||
XM_017018242.1 | XP_016873731.1 | 4 | 1994 | |||
XM_017018243.1 | XP_016873732.1 | 3 | 813 | |||
GenBank mRNA | AB070218.1 | - | 5 | 1340 | ||
AK075540.1 | - | 5 | 1113 | |||
BC113386.1 | - | 5 | 1321 | 8322438 | ||
BC113388.1 | - | 5 | 1321 | 8322440 | ||
JQ715612.1 | - | 5 | 1157 | |||
JQ715613.1 | - | 5 | 966 | |||
Y12692.1 | - | 5 | 1401 |