Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024815.3 | NP_079091.3 | 3 | 1275 | ||
XM_011544650.1 | XP_011542952.1 | 4 | 2006 | |||
GenBank mRNA | AK026147.1 | - | 2 | 1459 | ||
AK124446.1 | - | 4 | 2006 | |||
BC016902.1 | - | 3 | 1287 | 3914234 |