Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301098.1 | NP_001288027.1 | 4 | 1594 | ||
NM_001301099.1 | NP_001288028.1 | 4 | 1594 | |||
NM_014155.4 | NP_054874.3 | 4 | 1594 | |||
XM_005271521.4 | XP_005271578.1 | 4 | 1790 | |||
XM_005271522.4 | XP_005271579.1 | 4 | 1615 | |||
XM_006718825.3 | XP_006718888.1 | 4 | 1618 | |||
XM_006718826.3 | XP_006718889.1 | 4 | 1615 | |||
XM_011542789.2 | XP_011541091.1 | 4 | 1667 | |||
XM_017017623.1 | XP_016873112.1 | 4 | 1666 | |||
XM_017017624.1 | XP_016873113.1 | 4 | 1612 | |||
GenBank mRNA | AK024659.1 | - | 3 | 761 | ||
BC030580.1 | - | 4 | 1478 | 4823171 | ||
BC049375.1 | - | 4 | 1499 | 5271784 | ||
BC050723.1 | - | 3 | 1216 | 4445874 | ||
BC071729.1 | - | 4 | 1573 | 5163748 |