Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006544.3 | NP_006535.1 | 18 | 4802 | ||
XM_005267272.3 | XP_005267329.1 | 18 | 4887 | |||
GenBank mRNA | CB145811.1 | - | 1 | 38 |