Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SENP1 SIRNA Assays ›
Gene Symbol:
SENP1
Gene Name:
SUMO1/sentrin specific peptidase 1
Gene Aliases:
SuPr-2
Chromosome Location:
Chr.12: 48042893 - 48106308 on Build GRCh38
UniGene ID:
Hs.371957
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21696
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SENP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267594.1
NP_001254523.1
12, 13
1608
NM_001267595.1
NP_001254524.1
12, 13
1863
NR_051991.1
13, 14
2049
NR_051992.1
13, 14
2062
XM_006719361.3
XP_006719424.1
12, 13
1743
XM_006719362.2
XP_006719425.1
13, 14
1515
XM_011538244.2
XP_011536546.1
11, 12
1744
XM_011538245.2
XP_011536547.1
11, 12
1695
XM_017019229.1
XP_016874718.1
13, 14
1881
XM_017019230.1
XP_016874719.1
12, 13
1797
XM_017019231.1
XP_016874720.1
13, 14
1781
XM_017019232.1
XP_016874721.1
13, 14
1508
XM_017019233.1
XP_016874722.1
12, 13
1791
XM_017019234.1
XP_016874723.1
12, 13
1701
XM_017019235.1
XP_016874724.1
12, 13
1434
XM_017019236.1
XP_016874725.1
11, 12
1669
XM_017019237.1
XP_016874726.1
11, 12
1569
XM_017019238.1
XP_016874727.1
13, 14
1819
XM_017019239.1
XP_016874728.1
12, 13
1859
XM_017019240.1
XP_016874729.1
12, 13
1780
GenBank mRNA
AF149770.1
-
10, 11
1283
AK292060.1
-
12, 13
1379
AK312874.1
-
12, 13
1609
BC045639.1
-
12, 13
1679
5298667
BX537920.1
-
13, 14
2062
BX640784.1
-
13, 14
1893
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008765847, XM_008765848, XM_008765846, XM_008765849, XM_008765850, and others.
Panther Classification:
Molecular Function -
protease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
activation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of definitive erythrocyte differentiation
protein sumoylation
protein desumoylation
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
positive regulation of transcription from RNA polymerase II promoter
apoptotic signaling pathway
endopeptidase activity
protein binding
SUMO-specific protease activity
SUMO-specific endopeptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0