Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001414.3 | NP_001405.1 | 1, 2 | 233 | ||
GenBank mRNA | AK294815.1 | - | 1, 2 | 225 | ||
AL541292.3 | - | 1, 2 | 201 | |||
BC103763.1 | - | 1, 2 | 95 | 6148834 | ||
BC104188.1 | - | 24 | 40030987 | |||
BC104189.2 | - | 24 | 40030991 | |||
CR456831.1 | - | 14 | ||||
DA221985.1 | - | 1, 2 | 226 | |||
DA879094.1 | - | 1, 2 | 233 | |||
X95648.1 | - | 24 |