Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284306.1 | NP_001271235.1 | 6 | 1447 | ||
NM_001284307.1 | NP_001271236.1 | 8 | 1462 | |||
NM_022473.2 | NP_071918.1 | 7 | 3890 | |||
XM_005254591.2 | XP_005254648.1 | 10 | 3797 | |||
XM_005254592.2 | XP_005254649.1 | 10 | 3797 | |||
XM_017022483.1 | XP_016877972.1 | 9 | 3572 | |||
XM_017022484.1 | XP_016877973.1 | 7 | 3317 | |||
XM_017022485.1 | XP_016877974.1 | 8 | 1457 | |||
GenBank mRNA | AF205632.1 | - | 7 | 3863 | ||
AK127741.1 | - | 5 | 690 | |||
AK226132.1 | - | 8 | 1370 | |||
AK302735.1 | - | 9 | 1344 | |||
AL831983.1 | - | 6 | 1393 | |||
AL832014.1 | - | 6 | 1671 | |||
AL832352.1 | - | 6 | 1890 | |||
AL832367.1 | - | 5 | 602 | |||
AL832455.2 | - | 6 | 1803 | |||
AL833301.1 | - | 6 | 1901 | |||
BX648629.1 | - | 6 | 2995 |