Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284306.1 | NP_001271235.1 | 2 | 647 | ||
NM_001284307.1 | NP_001271236.1 | 4 | 662 | |||
NM_022473.2 | NP_071918.1 | 3 | 3090 | |||
XM_005254591.2 | XP_005254648.1 | 6 | 2997 | |||
XM_005254592.2 | XP_005254649.1 | 6 | 2997 | |||
XM_017022483.1 | XP_016877972.1 | 5 | 2772 | |||
XM_017022484.1 | XP_016877973.1 | 3 | 2517 | |||
XM_017022485.1 | XP_016877974.1 | 4 | 657 | |||
GenBank mRNA | AF205632.1 | - | 3 | 3063 | ||
AK057410.1 | - | 2 | 2071 | |||
AK091929.1 | - | 4 | 2826 | |||
AK226132.1 | - | 4 | 570 | |||
AK294529.1 | - | 3 | 2820 | |||
AK302735.1 | - | 5 | 544 | |||
AL831983.1 | - | 2 | 593 | |||
AL832014.1 | - | 2 | 871 | |||
AL832352.1 | - | 2 | 1090 | |||
AL832455.2 | - | 2 | 1004 | |||
AL833301.1 | - | 2 | 1101 | |||
BX648629.1 | - | 2 | 2195 |