Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENGASE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042573.2 | NP_001036038.1 | 3 | 267 | ||
XM_006722019.3 | XP_006722082.1 | 3 | 392 | |||
XM_017024961.1 | XP_016880450.1 | 2 | 109 | |||
GenBank mRNA | AF512564.1 | - | 3 | 259 | ||
AK025518.1 | - | 3 | 267 | |||
AK301115.1 | - | 3 | 265 |