Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031952.3 | NP_114158.2 | 5 | 798 | ||
NR_125327.1 | 5 | 798 | ||||
NR_125329.1 | 6 | 901 | ||||
NR_125330.1 | 4 | 570 | ||||
XM_011543664.1 | XP_011541966.1 | 5 | 754 | |||
XM_011543666.1 | XP_011541968.1 | 4 | 570 | |||
XM_011543667.2 | XP_011541969.1 | 4 | 976 | |||
XM_017009948.1 | XP_016865437.1 | 5 | 779 | |||
XM_017009949.1 | XP_016865438.1 | 10 | 1335 | |||
XM_017009950.1 | XP_016865439.1 | 9 | 1317 | |||
XM_017009951.1 | XP_016865440.1 | 4 | 706 | |||
GenBank mRNA | AK093225.1 | - | 4 | 570 | ||
AK292338.1 | - | 5 | 779 | |||
AY027526.1 | - | 5 | 747 | |||
BC032832.1 | - | 6 | 906 | 4827641 | ||
BC047333.1 | - | 5 | 784 | 5166271 | ||
DB027274.1 | - | 4 | 570 |