Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205319.1 | NP_001192248.1 | 7 | 792 | ||
NM_022463.4 | NP_071908.2 | 7 | 1135 | |||
XM_005256756.4 | XP_005256813.1 | 6 | 961 | |||
XM_005256758.3 | XP_005256815.1 | 7 | 1464 | |||
GenBank mRNA | AK022676.1 | - | 5 | 564 | ||
AK027451.1 | - | 6 | 688 | |||
AK297990.1 | - | 3 | 264 | |||
AK302073.1 | - | 7 | 794 | |||
AY189690.1 | - | 6 | 675 | |||
BC009327.2 | - | 7 | 729 | |||
BC063828.1 | - | 4 | 390 | |||
BC073845.1 | - | 5 | 545 |